missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse FIZZ1/RELM alpha Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB1523-100
This item is not returnable.
View return policy
Description
FIZZ1/RELM alpha Monoclonal specifically detects FIZZ1/RELM alpha in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| FIZZ1/RELM alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q9EP95 | |
| RETNLA | |
| E. coli-derived recombinant mouse FIZZ1/RELM alpha Asp24-Ser111 Accession # Q9EP95 | |
| 100 μg | |
| Primary | |
| Detects mouse FIZZ1/RELM alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) RELM beta or rmResistin is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 226033 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 1810019L16Rik, Fizz1, Fizz-1, HIMF, RELM alpha, RELMa, RELMalpha, RELM-alpha, RETNLA, Retnla resistin like alpha, RETNLB, Xcp2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 57262 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction