missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse CXCL2 (MIP-2) Recombinant Protein
E. coli Recombinant Protein
Brand: Invitrogen™ RP87878
Additional Details : Weight : 0.01000kg
Description
Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.Specifications
P10889 | |
Control, ELISA, Western Blot | |
20310 | |
Mouse CXCL2 (aa 28-100) | |
E. coli | |
RUO | |
C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 2; CINC-2 A; Cinc3; CINC-3; CXC; CXC motif chemokine; C-X-C motif chemokine 2; C-X-C motif chemokine ligand 2; CXCL; Cxcl2; cytokine-induced neutrophil chemoattractant 3; GRO b; gro beta; GRO ß; GRO2; GRO2 oncogene; GROB; GRObeta; Gro-beta; GRO-beta(5-73); GRO-beta-T; Growth-regulated protein beta; GROß; Hematopoietic synergistic factor; HSF; Macrophage inflammatory protein 2; macrophage inflammatory protein 2-alpha; melanoma growth stimulatory activity beta; MGSA beta; MGSA-b; MIP 2 A; MIP 2 alpha; MIP 2 A; Mip2; MIP-2; MIP2 a; MIP2 a; MIP2A; MIP-2 A; MIP2-alpha; SB-251353; Scyb; SCYB2; small inducible cytokine subfamily, member 2 | |
Mouse | |
Recombinant | |
>95% |
200 μg/mL | |
PBS with no preservative | |
7.8 kD | |
100 μg | |
-20°C | |
0.01 ng/ μg cytokine | |
Cxcl2 | |
Unconjugated | |
Liquid |