Learn More
Invitrogen™ Mouse CXCL2 (MIP-2) Recombinant Protein
E. coli Recombinant Protein
Brand:  Invitrogen™ RP87877
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 17616675
  • 2900.00 NOK / 25µg
Estimated Shipment: 03-06-2024
Description
Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.
Specifications
Specifications
P10889
Control, ELISA, Western Blot
20310
Mouse CXCL2 (aa 28-100)
E. coli
RUO
C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 2; CINC-2 A; Cinc3; CINC-3; CXC; CXC motif chemokine; C-X-C motif chemokine 2; C-X-C motif chemokine ligand 2; CXCL; Cxcl2; cytokine-induced neutrophil chemoattractant 3; GRO b; gro beta; GRO ß; GRO2; GRO2 oncogene; GROB; GRObeta; Gro-beta; GRO-beta(5-73); GRO-beta-T; Growth-regulated protein beta; GROß; Hematopoietic synergistic factor; HSF; Macrophage inflammatory protein 2; macrophage inflammatory protein 2-alpha; melanoma growth stimulatory activity beta; MGSA beta; MGSA-b; MIP 2 A; MIP 2 alpha; MIP 2 A; Mip2; MIP-2; MIP2 a; MIP2 a; MIP2A; MIP-2 A; MIP2-alpha; SB-251353; Scyb; SCYB2; small inducible cytokine subfamily, member 2
Mouse
Recombinant
>95%
200 μg/mL
PBS with no preservative
7.8 kD
25 μg
-20°C
0.01 ng/ μg cytokine
Cxcl2