missing translation for 'onlineSavingsMsg'
Learn More

Mouse CXCL16 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18767408
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
Product Code. Quantity unitSize
18767408 25 μg 25µg
18789397 100 μg 100µg
18775579 500 μg 500µg
3 options
This item is not returnable. View return policy

Product Code. 18767408

Brand: R&D Systems MAB503SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 4 publications

CXCL16 Monoclonal specifically detects CXCL16 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL16
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone 142417
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.15-0.6 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8BSU2
Gene Alias chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCL16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16
Gene Symbols CXCL16
Host Species Rat
Immunogen E. coli-derived recombinant mouse CXCL16 Asn27-Pro114 Accession # Q8BSU2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58191
Test Specificity Detects mouse CXCL16 in ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CXCL1, 2, 3, 5, 6, 7, 8, 9, 10, 11, 12, 13, 14, recombinant mouse CXCL1, 2, 6, 9, 10, 11, 12, 13, 14, 15, recombinant rat CXCL1, 2, 3, 5, or recombinant porcine CXCL8 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.