missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Mouse CD28 Antibody, R&D Systems™

Beskrivelse
CD28 Monoclonal specifically detects CD28 in Mouse samples. It is validated for Flow Cytometry, Agonist Activity, CyTOF-ready.
Tekniske data
Tekniske data
| Antigen | CD28 |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 794716 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Agonist Activity 0.07-0.42 ug/mL, CyTOF-ready |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P31041 |
| Gene Alias | CD28, CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 |
| Vis mere |
For Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?