missing translation for 'onlineSavingsMsg'
Learn More

Mouse CCL21/6Ckine Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18795488
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18795488 500 μg 500µg
18769437 25 μg 25µg
18791898 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18795488 Supplier R&D Systems Supplier No. MAB457500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 6 publications

CCL21/6Ckine Monoclonal specifically detects CCL21/6Ckine in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL21/6Ckine
Applications Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF, ELISA
Classification Monoclonal
Clone 59106
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 25 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P84444
Gene Alias 6Ckine, 6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, exodus-2, member 21, SCYA21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLC, SLCSecondary lymphoid-tissue chemokine, TCA-4
Gene Symbols Ccl21a
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCL21/6Ckine Ser24-Gly133 Accession # P84444
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6366
Test Specificity Detects mouse CCL21/6Ckine in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) 6Ckine, rhMIP-1 alpha , rhMIP-1 beta , recombinant mouse (rm) MIP-1 alpha , rmMIP-1 beta , rhMIP-1 delta , rmMIP-1 gamma , rmMIP-2, rhMIP-3 alpha , rmMIP-3 alpha , rhMIP-3 beta , rmMIP-3 beta , rhTeck, or rmTeck.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.