missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse CCL11 (Eotaxin) Recombinant Protein

E. coli Recombinant Protein

Brand:  Invitrogen™ RP87867

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 17616665

  • 2630.00 NOK / 25µg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
Specifications

Specifications

P48298
Control, ELISA, Western Blot
20292
Mouse Eotaxin (aa 24-97)
E. coli
RUO
C-C motif chemokine 11; C-C motif chemokine ligand 11; Ccl11; chemokine (C-C motif) ligand 11; eosinophil chemotactic protein; Eotaxin; Eotaxin 1; Eotaxin1; eotaxin-1; MGC22554; Scya11; small chemokine (C-C motif) ligand 11; small inducible cytokine A11; small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin); small inducible cytokine subfamily A11; small-inducible cytokine A11
Mouse
Recombinant
>95%
200 μg/mL
PBS with no preservative
8.4 kD
25 μg
-20°C
0.01 ng/ μg cytokine
CCL11
Unconjugated
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers