missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse CCL11 (Eotaxin) Recombinant Protein

Product Code. 17606665
Change view
Click to view available options
Quantity:
10 μg
100 μg
25 μg
Unit Size:
100µg
10µg
25µg
Product Code. Quantity unitSize
17606665 10 μg 10µg
17616665 25 μg 25µg
17626665 100 μg 100µg
3 options
This item is not returnable. View return policy

Product Code. 17606665

Brand: Invitrogen™ RP87866

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

E. coli Recombinant Protein

This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P48298
Concentration 200 μg/mL
For Use With (Application) Control, ELISA, Western Blot
Formulation PBS with no preservative
Gene ID (Entrez) 20292
Molecular Weight (g/mol) 8.4 kD
Name Mouse Eotaxin (aa 24-97)
Quantity 10 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration 0.01 ng/ μg cytokine
Gene Alias C-C motif chemokine 11; C-C motif chemokine ligand 11; Ccl11; chemokine (C-C motif) ligand 11; eosinophil chemotactic protein; Eotaxin; Eotaxin 1; Eotaxin1; eotaxin-1; MGC22554; Scya11; small chemokine (C-C motif) ligand 11; small inducible cytokine A11; small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin); small inducible cytokine subfamily A11; small-inducible cytokine A11
Gene Symbol CCL11
Specific Reactivity Mouse
Conjugate Unconjugated
Recombinant Recombinant
Form Liquid
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.