missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse BTN2A2/Butyrophilin 2A2 Protein, CF
Learn More About B7-related Butryophilins as Immune Modulators View Webinar
Brand: R&D Systems™ 8997-BT-050
This item is not returnable.
View return policy
Description
Measured by its ability to inhibit IL-2 secretion by mouse T cells in the presence of anti-CD3. The ED50 for this effect is 1-5 μg/mL.Specifications
| A4QPC6 | |
| 10385 | |
| 50 µg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| BTF2, BTF2BT2.2butyrophilin 2, BTN2, Butyrophilin 2, butyrophilin subfamily 2 member A2, butyrophilin, subfamily 2, member A2, FLJ41908 | |
| Btn2a2 | |
| Unconjugated | |
| Reconstitute at 200 µg/mL in PBS. |
| Lyophilized from a 0.2 µm filtered solution in PBS and EDTA. | |
| M.W. (Observed): 38-45 kDa, reducing conditions; M.W. (theoretical): 26 kDa | |
| Mouse myeloma cell line, NS0-derived mouse BTN2A2/Butyrophilin 2A2 protein Gln30-Trp248, with a C-terminal 6-His tag | |
| <0.10 EU / 1 µg of the protein by the LAL method. | |
| BTN2A2/Butyrophilin 2 | |
| Measured by its ability to inhibit IL-2 secretion by mouse T cells in the presence of anti-CD3. The ED50 for this effect is typically 1-5μg/mL. | |
| Yes |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction