missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse BD-3 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 250-41-5UG
Additional Details : Weight : 0.16500kg
Description
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].Specifications
Functional Assay | |
4.6 | |
5 μg | |
-20°C | |
<1 EU/μg | |
Mouse | |
Recombinant | |
Lyophilized |
protein with no preservative | |
Mouse BD-3 | |
E. coli | |
RUO | |
BD3; BD-3; beta defensin 3; beta defensin-3; beta-defensin 103; Beta-defensin 3; DEFB103; DEFB103A; DEFB103B; DEFB3; DEFB-3; defensin beta 103 A; defensin, beta 103; defensin, beta 103 A; defensin, beta 3; defensin-like protein; HBD3; hBD-3; HBP3; HBP-3 | |
Unconjugated | |
KKINNPVSCL RKGGRCWNRC IGNTRQIGSC GVPFLKCCKR K | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |