missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse BD-3 Recombinant Protein, PeproTech®

E. coli Recombinant Protein

Brand:  Gibco™ 250-41-500UG

Additional Details : Weight : 0.16500kg

 View more versions of this product

Product Code. 17870603

  • 17430.00 NOK / 500µg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].
Specifications

Specifications

Functional Assay
4.6
500 μg
-20°C
<1 EU/μg
Mouse
Recombinant
Lyophilized
protein with no preservative
Mouse BD-3
E. coli
RUO
BD3; BD-3; beta defensin 3; beta defensin-3; beta-defensin 103; Beta-defensin 3; DEFB103; DEFB103A; DEFB103B; DEFB3; DEFB-3; defensin beta 103 A; defensin, beta 103; defensin, beta 103 A; defensin, beta 3; defensin-like protein; HBD3; hBD-3; HBP3; HBP-3
Unconjugated
KKINNPVSCL RKGGRCWNRC IGNTRQIGSC GVPFLKCCKR K
≥ 98% by SDS-PAGE gel and HPLC analyses.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers