missing translation for 'onlineSavingsMsg'
Learn More

TP53, Mouse, Clone: FP3.2 [FPS392], Abnova™

Mouse monoclonal antibody raised against synthetic phosphopeptide of TP53.

Brand:  Abnova MAB0977.100ug

Product Code. 16069965

  • ONLINE EXCLUSIVE PRICE
    5525.00 NOK
    4808.55 NOK / 100µg
    Save 716.45 NOK
    12% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes tumor protein p53, which responds to diverse cellular stresses to regulate target genes that induce cell cycle arrest, apoptosis, senescence, DNA repair, or changes in metabolism. p53 protein is expressed at low level in normal cells and at a high level in a variety of transformed cell lines, where it's believed to contribute to transformation and malignancy. p53 is a DNA-binding protein containing transcription activation, DNA-binding, and oligomerization domains. It is postulated to bind to a p53-binding site and activate expression of downstream genes that inhibit growth and/or invasion, and thus function as a tumor suppressor. Mutants of p53 that frequently occur in a number of different human cancers fail to bind the consensus DNA binding site, and hence cause the loss of tumor suppressor activity. Alterations of this gene occur not only as somatic mutations in human malignancies, but also as germline mutations in some cancer-prone families with Li-Fraumeni syndrome. Multiple p53 variants due to alternative promoters and multiple alternative splicing have been found. These variants encode distinct isoforms, which can regulate p53 transcriptional activity. [provided by RefSeq]

Specifications

Specifications

TP53
Monoclonal
1 mg/mL
Mouse monoclonal antibody raised against synthetic phosphopeptide of TP53.
In PBS, pH 7.4 (0.09% sodium azide)
FLJ92943/LFS1/TRP53/p53
Mouse
100 μg
Primary
This antibody reacts with human p53 tumour suppressor protein phosphorylated at CKII site (Ser 392).
Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
IgG1
Immunohistochemistry, Western Blot
FP3.2 [FPS392]
Unconjugated
The optimal working dilution should be determined by the end user.
TP53
TP53
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S392 of human TP53.
RUO
7157
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.