missing translation for 'onlineSavingsMsg'
Learn More

SERPINA1 Mouse anti-Human, Monoclonal Antibody, Abnova™

Mouse monoclonal antibody raised against native SERPINA1.

Brand:  Abnova MAB6137.100ug

Product Code. 16056876

  • ONLINE EXCLUSIVE PRICE
    5270.00 NOK
    4582.87 NOK / 100µg
    Save 687.13 NOK
    13% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The protein encoded by this gene is secreted and is a serine protease inhibitor whose targets include elastase, plasmin, thrombin, trypsin, chymotrypsin, and plasminogen activator. Defects in this gene can cause emphysema or liver disease. Several transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Specifications

Specifications

SERPINA1
Monoclonal
Mouse monoclonal antibody raised against native SERPINA1.
In PBS, pH 7.4
A1A/A1AT/AAT/MGC23330/MGC9222/PI/PI1/PRO2275
Mouse
100 μg
Primary
These antibodies specifically recognize alpha-1-antitrypsin. They do not cross-react with alpha-1-antichymotrypsin.
Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
IgG2a
ELISA, Immunofluorescence
Unconjugated
The optimal working dilution should be determined by the end user.
SERPINA1
SERPINA1
Native purified SERPINA1 from human serum.
RUO
5265
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.