Learn More
Plasma Cell, Mouse, Clone: LIV3G11, Abnova™
Product Code. 16050676
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. Quantity unitSize
16050676 100 μg 100µg
1 options
Product Code. 16050676
Brand: Abnova MAB11268.100ug
Mouse monoclonal antibody raised against Plasma Cell.
Specifications
Antigen Plasma Cell
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone LIV3G11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Plasma Cell.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:400) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Host Species Mouse
Immunogen Pancreatic cancer related serum mucin isolated from human.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a