missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pdcd1, Mouse, Clone: 12A7D7, Abnova™
Mouse monoclonal antibody raised against partial recombinant Pdcd1.
Brand: Abnova MAB8089.100ug
This item is not returnable.
View return policy
Specifications
| Pdcd1 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| Pdcd1 | |
| Pdcd1 | |
| Recombinant protein corresponding to 150 amino acids of mouse Pdcd1. | |
| RUO | |
| 18566 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| 12A7D7 | |
| Mouse monoclonal antibody raised against partial recombinant Pdcd1. | |
| In PBS (0.02% sodium azide) | |
| Ly101/PD-1/Pdc1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur