Learn More
MAP2 Mouse anti-Human, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against partial recombinant MAP2.
Brand: Abnova MAB8746.100ug
Description
This gene encodes a protein that belongs to the microtubule-associated protein family. The proteins of this family are thought to be involved in microtubule assembly, which is an essential step in neurogenesis. The products of similar genes in rat and mouse are neuron-specific cytoskeletal proteins that are enriched in dentrites, implicating a role in determining and stabilizing dentritic shape during neuron development. A number of alternatively spliced variants encoding distinct isforms have been described. [provided by RefSeq
Specifications
| MAP2 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:100-1:2000) ELISA (1:5000-1:20000) The optimal working dilution should be determined by the end user. | |
| MAP2 | |
| DKFZp686I2148/MAP2A/MAP2B/MAP2C | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 4133 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against partial recombinant MAP2. | |
| In PBS, pH 7.4 (0.02% sodium azide, 1% BSA, 50% glycerol) | |
| NM_031846.2 | |
| MAP2 | |
| Recombinant protein corresponding to C-terminus residues of human MAP2. | |
| 100 μg | |
| Primary | |
| This antibody is specific to MAP2. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.