missing translation for 'onlineSavingsMsg'
Learn More

Infectious bronchitis virus (IBV), Mouse, Clone: IB95, Abnova™

Product Code. 16097426
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16097426 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16097426 Supplier Abnova Supplier No. MAB8524.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against Infectious bronchitis virus (IBV).

Specifications

Antigen Infectious bronchitis virus (IBV)
Applications ELISA, Western Blot
Classification Monoclonal
Clone IB95
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Infectious bronchitis virus (IBV).
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native semi-purified inactivated IB virus of Massachusetts type from infected chicken embryos.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Nucleoprotein of IBV.
Target Species Chicken
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.