Learn More
Mouse anti-Human IgG Fc Secondary Antibody (EM-07), Biotin, Novus Biologicals™
Product Code. 30232438 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30232438 0.1 mL 0.1mL
1 options
Product Code. 30232438
Brand: Novus Biologicals NBP145063B
Mouse Monoclonal Antibody
IgG Fc Monoclonal antibody specifically detects IgG Fc in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgG Fc
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone EM-07
Conjugate Biotin
Formulation PBS
Host Species Mouse
Immunogen Fusion protein of human IgG Fc fragment.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1