Learn More
Mouse anti-HO-1/HMOX1/HSP32, Clone: HO-1-1, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 14 publications
Brand:  Novus Biologicals NBP1-97507-0.2mg
Encompass
Product Code. 15709045
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
HO-1/HMOX1/HSP32 Monoclonal specifically detects HO-1/HMOX1/HSP32 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.