missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erythropoietin/EPO Mouse anti-Human, Clone: AE7A5, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2871-100
This item is not returnable.
View return policy
Description
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Human samples. It is validated for Western Blot, Immunoaffinity Purification.Specifications
| Erythropoietin/EPO | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2056 | |
| Human | |
| Supernatant |
| Western Blot, Affinity Purification | |
| AE7A5 | |
| Western Blot 1 ug/mL, Immunoaffinity Purification | |
| P01588 | |
| EPO | |
| Erythropoietin/EPO synthetic peptide analog, Ala28-Thr53, Accession # P01588 | |
| 100 μg | |
| Primary | |
| Recognizes an epitope within the first 26 amino acids at the NH2 terminus of human urinary and human recombinant EPO. Binds human and rat EPO but does not bind sheep, baboon, or dog EPO (1, 2). | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
| IgG2a |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?