Learn More
CAPZA2, Mouse, Clone: 40, Abnova™
Mouse monoclonal antibody raised against full-length recombinant CAPZA2.
Brand: Abnova MAB9969.100ug
Beskrivning
The protein encoded by this gene is a member of the F-actin capping protein alpha subunit family. It is the alpha subunit of the barbed-end actin binding protein Cap Z. By capping the barbed end of actin filaments, Cap Z regulates the growth of the actin filaments at the barbed end. [provided by RefSeq
Specifikationer
| CAPZA2 | |
| Monoclonal | |
| Unconjugated | |
| Western blot (1:500) The optimal working dilution should be determined by the end user. | |
| CAPZA2 | |
| CAPZA2 | |
| Recombinant protein corresponding to full length human CAPZA2. | |
| 100 μg | |
| Primary | |
| It can expression in SW480 whole cell lysate. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Western Blot | |
| 40 | |
| Mouse monoclonal antibody raised against full length recombinant CAPZA2. | |
| In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300) | |
| CAPPA2/CAPZ | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 830 | |
| Human | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.