Learn More
Invitrogen™ Motilin Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564415
Description
Immunogen sequence: AFVPIFTYGE LQRMQEKERN KGQKKSLSVW QRSGEEGPVD PAEPIREEEN Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 30%.
This gene encodes a small peptide hormone that is secreted by cells of the small intestine to regulate gastrointestinal contractions and motility. Proteolytic processing of the secreted protein produces the mature peptide and a byproduct referred to as motilin-associated peptide (MAP). Two transcript variants encoding different preproprotein isoforms but the same mature peptide have been found for this gene.
Specifications
| Motilin | |
| Polyclonal | |
| Unconjugated | |
| MLN | |
| gastrin-related protein; MAP; MGC138519; MLN; Motilin; Motilin-associated peptide; prepromotilin; promotilin; secreted protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4295 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P12872 | |
| MLN | |
| Recombinant protein corresponding to Human Motilin. Recombinant protein control fragment (Product #RP-104166). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.