Learn More
LTE Scientific™ Morrison Discard Box for Touchclave-R Autoclaves
Product Code. 12755405
Change view
Click to view available options
For Use With (Equipment):
For TCR/120/H
For TCR/120/V
For TCR/160/H
For TCR/60/H
For TCR/60/V
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12755405 For TCR/120/H Each
12795405 For TCR/120/V Each
12765405 For TCR/160/H Each
12745405 For TCR/60/H Each
12775405 For TCR/60/V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12755405 Supplier LTE Scientific™ Supplier No. TC/MOR/R2
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
For use with Touchclave-R Autoclaves
  • Choose discard box compatible with your Model
Specifications
For Use With (Equipment) For TCR/120/H
Product Type Discard Box