Learn More
Morpholine-d8 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Morpholine-d8 Hydrochloride |
| CAS | 1107650-56-5 |
| Molecular Formula | C4 D8 H N O . Cl H |
| Quantity | 250 mg |
| Synonym | Morpholine-2,2,3,3,5,5,6,6-d8 Hydrochloride (1:1), Morpholine-2,2,3,3,5,5,6,6-d8 HCl |
| InChI Formula | InChI=1S/C4H9NO.ClH/c1-3-6-4-2-5-1;/h5H,1-4H2;1H/i1D2,2D2,3D2,4D2; |
| SMILES | Cl.[2H]C1([2H])NC([2H])([2H])C([2H])([2H])OC1([2H])[2H] |
| IUPAC Name | 2,2,3,3,5,5,6,6-octadeuteriomorpholine;hydrochloride |
| Molecular Weight (g/mol) | 131.6306 |
| Formula Weight | 131.0953 |