Learn More
MorDalphos Pd G4, 98%, BLD Pharm™
Specifica
Specifica
| CAS | 2230788-66-4 |
| Molecular Formula | C44H57N2O4PPdS |
| MDL Number | MFCD30145864 |
| Quantity | 100 mg |
| SMILES | P(C1C=CC=CC=1N1CCOCC1)(C12CC3CC(CC(C3)C1)C2)(C12CC3CC(CC(C3)C1)C2)[Pd+2]1(N(C2=CC=CC=C2C2=CC=CC=[C-]12)C)[O-]S(=O)(=O)C |
| Molecular Weight (g/mol) | 847.39 |
| Percent Purity | 98% |
| Chemical Name or Material | MorDalphos Pd G4 |