Learn More
Mono-POC Isopropyl Tenofovir (Mixture of Diastereomers), TRC
Product Code. 30469911
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469911 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30469911 Supplier Toronto Research Chemicals Supplier No. TRCM5669705MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Mono-POC Isopropyl Tenofovir (mixture of diastereomers)
CAS 1246812-40-7
Molecular Formula C17 H28 N5 O7 P
Quantity 5 mg
Synonym 2,4,7-Trioxa-5-phosphanonanoic acid, 9-(6-amino-9H-purin-9-yl)-8-methyl-5-(1-methylethoxy)-, 1-methylethyl ester, 5-oxide, (8R)-, [[[(2R)-3-(6-Amino-9H-purin-9-yl)prop-2-yloxy]methyl]-1-methylethoxyphosphoryl]oxymethyl 1-Methylethyl Carbonate, [[[(2R)-1-(6-Amino-9H-purin-9-yl)propan-2-yloxy]methyl](isopropyloxy)phosphoryl]oxymethyl Isopropyl Carbonate
InChI Formula InChI=1S/C17H28N5O7P/c1-11(2)28-17(23)25-9-27-30(24,29-12(3)4)10-26-13(5)6-22-8-21-14-15(18)19-7-20-16(14)22/h7-8,11-13H,6,9-10H2,1-5H3,(H2,18,19,20)/t13-,30?/m1/s1
SMILES CC(C)OC(=O)OCOP(=O)(CO[C@H](C)Cn1cnc2c(N)ncnc12)OC(C)C
IUPAC Name [[(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-propan-2-yloxyphosphoryl]oxymethyl propan-2-yl carbonate
Molecular Weight (g/mol) 445.41
Formula Weight 445.1726