Learn More
Molybdovanadic Reagent, Chem Lab™
Product Code. 11947443
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11947443 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11947443 Supplier Chem-Lab Supplier No. CL07.2201.0100
Specifications
Color Yellow
Boiling Point 100°C
Melting Point 0°C
Composition 1g NH4VO3 + 40g (NH4)6Mo7O24.4H2O / l 13% HNO3
Density 1.1g/mL
Solubility Information Water Soluble
Odor Odorless
pH <1
Quantity 100 mL
Physical Form Liquid
compliance-icons
Product Identifier
  • Molybdovanadic reagent
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Molybdovanadic