missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum Standard Solution, WTW™

Catalog Number 15297259
Click to view available options
Quantity:
10 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Physical Form Liquid
Solution Contains Molybdenum
Traceability to NIST Yes
Color Red/Violet
Quantity 10 mL
Solubility Soluble in water
compliance-icons
Product Identifier
  • molydenum cell test method
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : N-cetylpyridinium chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'