Learn More
Molybdenum Standard Metal Solution 1000ppm, for ICP, Solution NH3 0.1M, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
| Packaging | HDPE plastic bottle |
| Physical Form | Liquid |
| MDL Number | 3465 |
| Color | Colorless |
| Quantity | 500 mL |