Learn More
Agencinox™ Molybdene Stainless Steel Beakers with Spout
Product Code. p-8008855
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
11701903 100 mL Each
11711903 250 mL Each
11721903 500 mL Each
11751903 1000 mL Each
11761903 2000 mL Each
11771903 3000 mL Each
6 options
Product Code. 11701903
Brand: Agencinox™ 2010B
Only null left
Add to basket
Add to basket
Use these beakers for a variety of general laboratory applications. Agencinox™ Molybdenum Stainless Steel Beakers ensure the safe transfer of liquids.
  • Stainless steel Molybdenum construction
  • High chemical resistance
  • Available in a variety of sizes
Specifications
Diameter (Metric) Outer 50 mm
Capacity (Metric) 100 mL
Material Stainless Steel (Molybdenum)
Height (Metric) 60 mm
Type Beaker