missing translation for 'onlineSavingsMsg'
Learn More

Molindone hydrochloride, MedChemExpress

Product Code. p-200099890
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16458648

Brand: Medchem Express HYB1017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Molindone hydrochloride (EN-1733A) is a therapeutic antipsychotic, used in the treatment of schizophrenia, works by blocking the effects of dopamine in the brain, leading to diminished psychoses.

Molindone hydrochloride (EN-1733A) is a therapeutic antipsychotic, used in the treatment of schizophrenia, works by blocking the effects of dopamine in the brain, leading to diminished psychoses.

Specifications

Chemical Name or Material Molindone hydrochloride
Physical Form Solid
CAS 15622-65-8
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H25ClN2O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym EN-1733A
Solubility Information H2O : 100 mg/mL (319.66 mM; Need ultrasonic) ∣DMSO : 17.5 mg/mL (55.94 mM; Need ultrasonic and warming)
SMILES O=C1C2=C(NC(C)=C2CC)CCC1CN3CCOCC3.[H]Cl
Molecular Weight (g/mol) 312.83
Formula Weight 312.83
Percent Purity 99.5%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.