Learn More
Invitrogen™ X-Rhod-1, AM, cell permeant - Special Packaging
Product Code. 11500896
Click to view available options
Quantity:
10 x 50 μg
Unit Size:
500µg
Product Code. 11500896
Brand: Invitrogen™ X14210
Labeled calcium indicators that exhibit an increase in fluorescence upon binding Ca2+
  • Uses in many calcium signaling investigations, including measuring Ca2+ in cells and tissues
  • Cells may be loaded with the AM ester forms of these calcium indicators by adding the dissolved indicator directly to dishes containing cultured cells
  • Fluorescence signal from these cells is generally measured using fluorescence microscopy
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
  • In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+
  • Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelator
  • Label (Ex/Em of Ca2+–bound form): X-rhod-1 (580/602nm)
  • Fluorescence intensity increase upon binding Ca2+: >100 fold
  • Kd for Ca2+ in the absence of Mg2+, in buffer: approximately 700nM
Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Fluorescence Microscope
Product Type Dye
Dye Type Fluorescent Dye-Based
Quantity 10 x 50 μg
Shipping Condition Room Temperature