Learn More
Invitrogen™ Tubulin Tracker™ Green (Oregon Green™ 488 Taxol, Bis-Acetate), for live-cell imaging
Product Code. 10377872
Change view
Click to view available options
Quantity:
1 set
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10377872 1 set Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10377872 Supplier Invitrogen™ Supplier No. T34075
For live-cell imaging
Tubulin Tracker Green (Oregon Green 488 Taxol, bis-acetate) provides green-fluorescent staining of polymerized tubulin in live cells. Tubulin Tracker Green is an uncharged, nonfluorescent compound that easily passes through the plasma membrane of live cells. Once inside the cell, the lipophilic blocking group is cleaved by non-specific esterases, resulting in a green-fluorescent, charged form.
Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cytoskeleton, Organelle Tracing
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Dye Type Oregon Green™ 488
Quantity 1 set
Product Line Oregon Green
Shipping Condition Room Temperature
Label Type Classic Dyes
Product Type Tubulin Tracker
SubCellular Localization Cytoskeleton, Tubulin