Learn More
Invitrogen™ PBFI, AM, cell permeant - Special Packaging
Product Code. 11599196
Change view
Click to view available options
Quantity:
20 x 50 μg
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11599196 20 x 50 μg Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11599196 Supplier Invitrogen™ Supplier No. P1267MP
  • ONLINE EXCLUSIVE PRICE
    11210.00 NOK
    9752.70 NOK / Pack of 20
    Save 1457.30 NOK
    13% Off
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
PBFI is potassium-sensitive molecule used measure intracellular K+ fluxes in animal cells and in plant cells and vacuoles
  • Although selectivity of PBFI for K+ is less than that of calcium indicators such as fura-2, it is sufficient for detection of physiological concentrations of K+ in presence of other monovalent cations
  • Spectral response of PBFI upon ion binding permit excitation ratio measurements, and this indicator can be used with same optical filters and equipment used for fura-2
  • Label (Ex/Em): PBFI (approximately 340,380/500nm)
  • Lyophilized product may be dissolved in distilled water or aqueous buffer for use
  • Product is typically loaded into cells by diffusion from patch pipet for correlated fluorescence imaging and electrophysiological recording
  • Kd of PBFI for K+ is strongly dependent on whether Na+ is present, with value of 5.1mm in absence of Na+ and 44mm in solutions with combined Na+ and K+ concentration of 135 mm (which approximates physiological ionic strength)
  • Although PBFI is only 1.5-fold more selective for K+ than for Na+, this selectivity is often sufficient because intracellular K+ concentrations are normally about 10 times higher than Na+ concentrations
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Cell-Based Ion Channel Assays, Drug Discovery and Development, Ion Channel Biology, Ionic Homeostasis and Signaling, Sodium and Potassium Detection, Target and Lead Identification and Validation
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Fluorescence Microscope
Product Type PBFI AM
Dye Type Potassium Indicator
Quantity 20 x 50 μg
Shipping Condition Room Temperature