Learn More
Invitrogen™ MANT-GMPPNP (2'-(or-3')-O-(N-Methylanthraniloyl)-β:γ-Imidoguanosine 5'-Triphosphate, Trisodium Salt)
Description
The nucleotide analog MANT GMPPNP is modified on the ribose moiety. The compact mature of the MANT fluorophore and its attachment position results in nucleotide analogs that induce minimal perturbation of nucleotide-protein interactions. Because MANT fluorescence is sensitive to the environment of the fluorophore, nucleotide-protein interactions may be directly detectable. MANT nucleotides are valuable probes of the structure and enzymatic activity of nucleotide-binding proteins.
Drug Discovery and Development, Enzyme and Protein Activity Assays, Phosphatase Activity, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation
Order Info
Shipping Condition: Wet ice
Specifications
Specifications
| Product Type | MANT-GMPPNP |
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Shipping Condition | Wet Ice |
| Concentration | 5 mM |
| Label or Dye | MANT (N-Methylanthraniloyl) |
| Quantity | 50 μL |