Learn More
Invitrogen™ Ethidium Monoazide Bromide (EMA)
Description
After photolysis, Ethidium monoazide binds covalently to nucleic acids both in solution and in cells that have compromised membranes. The fluorescence of ethidium monoazide is weak, but the intensity increases ∽15-fold on binding to DNA with excitation/emission maxima of ∽504/600nm.
Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function, Nucleus, Nucleoli & Nuclear Envelope, Organelle Tracing
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Color | Orange |
| Content And Storage | Store in freezer at -5°C to -30°C and protect from light. |
| Excitation Wavelength Range | 504 nm |
| Dye Type | Cell-Impermeant |
| For Use With (Application) | Cell staining assays |
| For Use With (Equipment) | Flow Cytometer |
| Quantity | 5 mg |
| Detection Method | Fluorescence |
| Description | Ethidium Monoazide Bromide (EMA) |
| Emission | 600 nm |
| Show More |