Learn More
Invitrogen™ BODIPY™ FL C5-Lactosylceramide complexed to BSA
Product Code. 10730574
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10730574 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10730574 Supplier Invitrogen™ Supplier No. B34402
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 27-10-2026
The green-fluorescent BODIPY™ FL C5 lactosylceramide complexed to BSA can be used in sphingolipid transport and metabolism mechanisms.
The green-fluorescent BODIPY™ FL C5 lactosylceramide complexed to BSA can be used in sphingolipid transport and metabolism mechanisms. Complexing fluorescent lipids with bovine serum albuin (BSA) facilitates cell labeling by eliminating the need for organic solvents to dissolve the lipophilic probe—the BSA-complexed probe can be directly dissolved in water.
Consult user Manual for solubility instructions.
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sphingolipids
Quantity 1 mg
Physical Form Solid
Product Line BODIPY
Excitation/Emission 505/511 nm