Learn More
Invitrogen™ BODIPY™ FL C5-Ganglioside GM1
Description
Colocalization of fluorescent cholera toxin B conjugates and the green-fluorescent BODIPY FL C5-ganglioside GM1 observed by fluorescence microscopy provides a direct indication of the association of these molecules in lipid rafts.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Chemical Name or Material | Sphingolipids |
| Quantity | 25 μg |
| Physical Form | Solid |
| Product Line | BODIPY |