Learn More
Invitrogen™ BODIPY™ FL C12 (4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoic Acid)
Product Code. 10615583
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10615583 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10615583 Supplier Invitrogen™ Supplier No. D3822
Edge
The green fluorescent fatty acid, BODIPY™ FL C12 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids.
The green fluorescent fatty acid, BODIPY™ FL C12 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids. Unlike fluorophores like pyrene, DPH and NBD, BODIPY™ dyes are relatively environment insensitive and is fluorescent in both aqueous and lipid environments.
Dissolve in high-quality anhydrous DMSO for stock concentrations of 1 to 10 mM.
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Fatty Acids
Quantity 1 mg
Molecular Weight (g/mol) 418.33
Physical Form Solid
Product Line BODIPY