Learn More
Corning™ Molecular Biology Reagents
Description
- Certificate of Analysis available
- Product can be diluted to obtain the desired concentration
Order Info
Shipped at ambient temperature
Specifications
Specifications
| Chemical Name or Material | Tris Hydrochloride Buffer |
| Concentration | 1 M |
| pH | 8 ±0.1 |
| Physical Form | Liquid |
| Quantity | 6 x 1 L |
| Sterility | Sterile |