missing translation for 'onlineSavingsMsg'
Learn More

MOK Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 23926-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16892525

  • 3468.15 NOK / 150µL
Estimated Shipment: 19-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Able to phosphorylate several exogenous substrates and to undergo autophosphorylation.
Specifications

Specifications

MOK
Polyclonal
Unconjugated
MOK
MOK, MOK protein kinase, RAGE, RAGE 1, RAGE1, renal tumor antigen, Renal tumor antigen 1
Rabbit
Antigen Affinity Chromatography
RUO
5891
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Flow Cytometry
0.46 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UQ07
MOK
MOK Fusion Protein Ag20849
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers