Learn More
Moguisteine, TRC
Product Code. 30476378
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476378 100 mg 100mg
30474938 1 g 1g
30465830 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30476378 Supplier Toronto Research Chemicals Supplier No. TRCM486450100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 119637-67-1
Molecular Formula C16H21NO5S
Molecular Weight (g/mol) 339.41
Synonym (+/-)-2-[(2-Methoxyphenoxy)methyl]-beta-oxo-3-thiazolidinepropanoic Acid Ethyl Ester
IUPAC Name ethyl 3-[2-[(2-methoxyphenoxy)methyl]-1,3-thiazolidin-3-yl]-3-oxopropanoate
SMILES CCOC(=O)CC(=O)N1CCSC1COc2ccccc2OC
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C16H21NO5S/c1-3-21-16(19)10-14(18)17-8-9-23-15(17)11-22-13-7-5-4-6-12(13)20-2/h4-7,15H,3,8-11H2,1-2H3
Formula Weight 339.114
Chemical Name or Material Moguisteine