Learn More
Mogrol, MedChemExpress
Product Code. 16489278
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
Product Code. 16489278
Brand: Medchem Express HYN2312
MedChemExpress Mogrol is a biometabolite of mogrosides, and acts via inhibition of the ERK1/2 and STAT3 pathways, or reducing CREB activation and activating AMPK signaling.
Mogrol is a biometabolite of mogrosides, and acts via inhibition of the ERK1/2 and STAT3 pathways, or reducing CREB activation and activating AMPK signaling.
Specifications
Chemical Name or Material Mogrol
CAS 88930-15-8
Color White
Purity Grade Notes Research
Molecular Formula C30H52O4
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (104.88 mM; Need ultrasonic)
SMILES CC1(C)[C@@H](O)CC[C@@]2([H])[C@]3(C)[C@H](O)C[C@]4(C)[C@@H]([C@H](C)CC[C@@H](O)C(C)(O)C)CC[C@](C)4[C@]3([H])CC=C12
Molecular Weight (g/mol) 476.73
Formula Weight 476.73
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.