Learn More
BINDER™ Modular Plug Distributor
Shop All BINDER Products
Click to view available options
Material:
Plastic
Unit Size:
Each
Description
- For use with a wide range of chambers and incubators
Specifications
Specifications
| Description | RS 422 interface, accessories - modular plug distributor for 10 RS 422 interfaces |
| For Use With (Equipment) | Multi Management Software APT-COM™ |
| Material | Plastic |
| Type | Modular Plug Distributor |
| For Use With (Application) | RSS Interfacing |