Learn More
Ansell Model 400 - AlphaTec™ 3000 Overshoes Ultrasonically Welded and Taped
Product Code. 30690002 Shop All Ansell Products
Change view
Click to view available options
Size:
Universal
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30690002 Universal Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30690002 Supplier Ansell Supplier No. Y30T400
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 18-11-2026
Durable, comfortable multi-layer chemical barrier.
  • One of the lightest and most comfortable chemical protective garments, this durable multi-layer fabric provides an extremely effective barrier against both inorganic chemicals and biological hazards.
  • Provides a strong liquid and particle barrier
  • Overshoes, elasticated opening
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Material Multi-layer Barrier Laminate Fabric
Size Universal
Waterproof Water Repellent
Product Type Overshoe