Learn More
Köttermann Mobile Cabinet with Two Doors
Product Code. 17352109
Change view
Click to view available options
No. of Drawers:
1
2
None
Dimensions (D x W x H):
516 x 900 x 590 mm
516 x 900 x 740 mm
516 x 1200 x 590 mm
516 x 1200 x 740 mm
Unit Size:
Each
Product Code. No. of Drawers Dimensions (D x W x H) unitSize
17352109 None 516 x 1200 x 590 mm Each
17312129 None 516 x 900 x 590 mm Each
17332129 1 516 x 900 x 740 mm Each
17342129 2 516 x 900 x 740 mm Each
17372109 1 516 x 1200 x 740 mm Each
17382109 2 516 x 1200 x 740 mm Each
6 options
Product Code. 17352109
Brand: Köttermann 31500095
This item requires a price update prior to being ordered.
Request a Price for this item here.
This Mobile Cabinet with Two Doors offers versatile storage space with two doors to protect stored items.
  • Versatile storage space
  • Mobile with lockable wheels
  • Features two doors to protect stored items
  • Many sizes and configurations available
Specifications
Color White
Cabinet Type Mobile
Dimensions (D x W x H) 516 x 1200 x 590 mm
Door Type Hinged
Finish Epoxy Coated
Frame Material Steel
Hinge Side Left and Right
No. of Doors 2
No. of Drawers None
No. of Shelves None
Warranty None
Wheel Type Plastic with Brakes
With Casters Yes
Material Steel