Learn More
Köttermann Mobile Cabinet with One Lockable Door
Product Code. 17332119
Change view
Click to view available options
Hinge Side:
Right
No. of Drawers:
1
None
No. of Shelves:
1
None
Dimensions (D x W x H):
516 x 450 x 590 mm
516 x 450 x 740 mm
516 x 600 x 590 mm
516 x 600 x 740 mm
Unit Size:
Each
Product Code. Hinge Side No. of Drawers No. of Shelves Dimensions (D x W x H) unitSize
17332119 Right None None 516 x 450 x 590 mm Each
17372119 Right None 1 516 x 450 x 740 mm Each
17392119 Right 1 None 516 x 450 x 740 mm Each
17382099 Right None None 516 x 600 x 590 mm Each
17322109 Right None 1 516 x 600 x 740 mm Each
17332109 Right 1 None 516 x 600 x 740 mm Each
6 options
Product Code. 17332119
Brand: Köttermann 31500113
This item requires a price update prior to being ordered.
Request a Price for this item here.
This Mobile Cabinet with One Lockable Door offers versatile storage space with a single door to protect stored items.
  • Versatile storage space
  • Mobile with lockable wheels
  • Features one door with locking mechanism to protect stored items
  • Many sizes and configurations available
Specifications
Color White
Cabinet Type Mobile
Dimensions (D x W x H) 516 x 450 x 590 mm
Door Type Hinged
Finish Epoxy Coated
Frame Material Steel
Hinge Side Right
Locking Mechanism Yes
No. of Doors 1
No. of Drawers None
No. of Shelves None
Warranty None
Wheel Type Plastic with Brakes
With Casters Yes
Material Steel