Learn More
MNI-caged-L-glutamate, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | MNI-caged-L-glutamate |
| CAS | 295325-62-1 |
| Molecular Formula | C14 H17 N3 O6 |
| Quantity | 50 mg |
| Synonym | (αS)-Amino-2,3-dihydro-4-methoxy-7-nitro-Δ-oxo-1H-indole-1-pentanoic Acid |
| InChI Formula | InChI=1S/C14H17N3O6/c1-23-11-4-3-10(17(21)22)13-8(11)6-7-16(13)12(18)5-2-9(15)14(19)20/h3-4,9H,2,5-7,15H2,1H3,(H,19,20)/t9-/m0/s1 |
| SMILES | COc1ccc(c2N(CCc12)C(=O)CC[C@H](N)C(=O)O)[N+](=O)[O-] |
| IUPAC Name | (2S)-2-amino-5-(4-methoxy-7-nitro-indolin-1-yl)-5-oxo-pentanoic acid |
| Molecular Weight (g/mol) | 323.3 |
| Formula Weight | 323.1117 |