Learn More
Macherey-Nagel™ MN OPTIMA™ Amine Test Mixture in Ethanol
Product Code. 10115041 Shop All Macherey-Nagel Products
Change view
Click to view available options
Volume (Metric):
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10115041 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10115041 Supplier Macherey-Nagel™ Supplier No. 722317
  • ONLINE EXCLUSIVE PRICE
    510.00 NOK
    433.58 NOK / 1mL
    Save 76.42 NOK
    14% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Specifications
For Use With (Equipment) For GC Capillary Columns
Includes 0.2 % Diisobutylamine, 1 % Diethanolamine, 0.2 % 2,6-Dimethylaniline, 0.2 % o-Propanolpyridine, 0.2 % Dicyclohexylamine, 0.2 % Dibenzylamine
Volume (Metric) 1 mL
Product Type MN OPTIMA Amine Test Mixture in Ethanol
compliance-icons
Product Identifier
  • Test mixture OPTIMA 5 Amine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Supplemental information
  • MIXTURE LIST-Contain: ethanol