missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The Mmp9 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of mouse Mmp9. This antibody recognizes mouse antigen. The Mmp9 antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
Specifications
| Antigen | Mmp9 |
| Applications | Western Blot, ELISA |
| Clone | 5A1 |
| Concentration | 1000 μg/mL |
| Gene Symbols | AW743869, B/MMP9, Clg4b, matrix metallopeptidase 9, MMP 9, Mmp9, pro MMP 9 |
| Host Species | Rabbit |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 17395 |
| Target Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?